Assay Details
| Gene Symbol: Ifi204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008329.2 | NP_032355.2 | 2 - 3 | 388 | 93 | |
| GenBank mRNA | AK150522.1 | - | 2 - 3 | 389 | 93 | |
| BC010546.1 | - | 345 | 4018506 | 93 | ||
| CT010351.1 | - | 259 | 93 | |||
| M31419.1 | - | 386 | 93 | |||
| Gene Symbol: Mnda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033450.4 | NP_001028622.1 | 2 - 3 | 512 | 93 | |
| GenBank mRNA | AK152621.1 | - | 1 - 2 | 46 | 93 | |
| AK152683.1 | - | 2 - 3 | 380 | 93 | ||
| AK152786.1 | - | 2 - 3 | 389 | 93 | ||
| AK166217.1 | - | 2 - 3 | 512 | 93 | ||
| BC132314.1 | - | 2 - 3 | 333 | 40130591 | 93 | |
| BC132316.1 | - | 2 - 3 | 333 | 40130593 | 93 | |
| M74123.1 | - | 2 - 3 | 367 | 93 | ||
| S62227.1 | - | 2 - 3 | 367 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK152621.1 transcripts.
Set Membership: |
|

Ifi204
Genomic Map