Assay Details
| Gene Symbol: Tpmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016785.2 | NP_058065.2 | 1 - 2 | 111 | 114 | |
| GenBank mRNA | AF046887.1 | - | 1 - 2 | 97 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF046887.1, NM_016785.2 transcripts.
Set Membership: |
|

Tpmt
Genomic Map