Assay Details
| Gene Symbol: Ruvbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019685.2 | NP_062659.1 | 5 - 6 | 712 | 109 | |
| GenBank mRNA | AF100694.1 | - | 5 - 6 | 751 | 109 | |
| AK012170.1 | - | 5 - 6 | 660 | 109 | ||
| AK132783.1 | - | 5 - 6 | 694 | 109 | ||
| AK146258.1 | - | 5 - 6 | 704 | 109 | ||
| AK146374.1 | - | 5 - 6 | 705 | 109 | ||
| AK156078.1 | - | 5 - 6 | 682 | 109 | ||
| BC004718.1 | - | 5 - 6 | 678 | 3582426 | 109 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ruvbl1
Genomic Map