| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700022379.mm.1 |
NM_019940.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022379.mm.1 |
NM_019940.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022379.mm.1 |
TC0700002591.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700022381.mm.1 |
NM_019940.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022381.mm.1 |
NM_019940.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022381.mm.1 |
TC0700002591.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700022373.mm.1 |
NM_019940.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022373.mm.1 |
NM_019940.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022373.mm.1 |
TC0700002591.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700022378.mm.1 |
NM_019940.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022378.mm.1 |
NM_019940.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022378.mm.1 |
TC0700002591.mm.1 |
MTA TC |