Assay Details
| Gene Symbol: Ifnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024673.2 | NP_001019844.2 | 1 - 2 | 172 | 80 | |
| GenBank mRNA | AY869695.1 | - | 1 - 2 | 172 | 80 | |
| BC150796.1 | - | 1 - 2 | 267 | 9087707 | 80 | |
| Gene Symbol: Ifnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY869696.1 | - | 1 - 2 | 172 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ifnl2
Genomic Map