Assay Details
| Gene Symbol: Ifnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024673.2 | NP_001019844.2 | 3 - 4 | 398 | 102 | |
| GenBank mRNA | AY869695.1 | - | 3 - 4 | 398 | 102 | |
| BC150796.1 | - | 3 - 4 | 493 | 9087707 | 102 | |
| Gene Symbol: Ifnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177396.1 | NP_796370.1 | 3 - 4 | 398 | 102 | |
| GenBank mRNA | AY184375.1 | - | 3 - 4 | 398 | 102 | |
| AY869696.1 | - | 3 - 4 | 398 | 102 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ifnl2
Genomic Map