Assay Details
| Gene Symbol: Ifnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024673.2 | NP_001019844.2 | 4 - 5 | 480 | 89 | |
| GenBank mRNA | AY869695.1 | - | 4 - 5 | 480 | 89 | |
| BC150796.1 | - | 4 - 5 | 575 | 9087707 | 89 | |
| Gene Symbol: Ifnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY869696.1 | - | 4 - 5 | 480 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022265 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05912751 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05912745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06455853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06112858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05912725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06445018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06178385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06001929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05912743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06159327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06112867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05912751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05912734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06399840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022211 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10561477 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551527 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561476 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561475 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10551526 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561474 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700023662.mm.1 | NM_001024673.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023662.mm.1 | NM_001024673.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023662.mm.1 | TC0700002699.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700023661.mm.1 | TC0700002699.mm.1 | MTA TC |

Ifnl2
Genomic Map