Assay Details
| Gene Symbol: AW554918 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033532.3 | NP_001028704.2 | 7 - 8 | 1259 | 109 | |
| GenBank mRNA | AK220161.1 | - | 1 - 2 | 81 | 109 | |
| BC066089.1 | - | 2 - 3 | 774 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

AW554918
Genomic Map