Assay Details
| Gene Symbol: Gpr179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081220.1 | NP_001074689.1 | 10 - 11 | 2095 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962851 | NM_001081220.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768801 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77069 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640555 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 950947 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 716301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 716564 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166383 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574519 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340963 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17647 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63096 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144939 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962851 | NM_001081220.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144939 | - | - |
| Whole Mouse Genome G4122A | A_51_P188227 | AK030625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188227 | AK038894.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P442126 | NM_001081220.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P442126 | BC058657.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144939 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09458435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28049991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28050022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28049953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09535041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09279851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28049940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09015285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09287634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28049942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09163883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28049952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09279852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09223918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28049948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28049943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09108071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28049945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09223917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09279861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28050093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09412569 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10390420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390412 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390418 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390409 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390408 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390410 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390413 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390414 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390419 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390415 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390411 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390417 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390416 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459268_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10390407 | - | - |
| Mouse Genome 430 2.0 Array | 1459268_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459268_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217143_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217143_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100033483.mm.1 | NM_001081220.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100033483.mm.1 | NM_001081220.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100033483.mm.1 | TC1100003644.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1459268_at | - | - |
| Mouse Expression Set 430 | 1459268_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459268_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459268_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 380830 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2756275 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2756275 | - | - |

Gpr179
Genomic Map