Assay Details
| Gene Symbol: Gpr179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081220.1 | NP_001074689.1 | 1 - 2 | 852 | 94 | |
| GenBank mRNA | AK030625.1 | - | 1 - 2 | 856 | 94 | |
| AK038894.1 | - | 1 - 2 | 846 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr179
Genomic Map