Assay Details
| Gene Symbol: Spata31d1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083890.2 | NP_001077359.2 | 2 - 3 | 255 | 85 | |
| GenBank mRNA | AK133350.1 | - | 2 - 3 | 256 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spata31d1c
Genomic Map