Assay Details
| Gene Symbol: Spata31d1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083890.2 | NP_001077359.2 | 3 - 4 | 307 | 98 | |
| GenBank mRNA | AK133350.1 | - | 3 - 4 | 308 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987879 | NM_001083890.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987879 | AK133350.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 895213 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717946 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 118610 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42710 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 287720 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 70664 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987879 | NM_001083890.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987879 | AK133350.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07902388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29101534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11089592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101502 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10538841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10538844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10538846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11122345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10538835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11089598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29101508 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10405845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405844 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405846 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10405842 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300005195.mm.1 | NM_001083890.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005195.mm.1 | NM_001083890.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300005195.mm.1 | TC1300000785.mm.1 | MTA TC |

Spata31d1c
Genomic Map