Assay Details
| Gene Symbol: Myo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103171.1 | NP_001096641.1 | 20 - 21 | 5604 | 76 | |
| NM_010862.2 | NP_034992.2 | 20 - 21 | 5604 | 76 | ||
| NM_182698.2 | NP_874357.2 | 19 - 20 | 1842 | 76 | ||
| GenBank mRNA | AB014510.1 | - | 16 - 17 | 1607 | 76 | |
| AF053130.1 | - | 18 - 19 | 1810 | 76 | ||
| AF144095.1 | - | 20 - 21 | 5604 | 76 | ||
| AY331132.1 | - | 20 - 21 | 5604 | 76 | ||
| AY331133.1 | - | 19 - 20 | 1842 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myo15
Genomic Map