| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100006020.mm.1 |
NM_001103171.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006020.mm.1 |
NM_001103171.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006020.mm.1 |
NM_010862.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006020.mm.1 |
NM_010862.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006020.mm.1 |
NM_182698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006020.mm.1 |
NM_182698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006020.mm.1 |
TC1100000755.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006019.mm.1 |
NM_001103171.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006019.mm.1 |
NM_001103171.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006019.mm.1 |
NM_010862.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006019.mm.1 |
NM_010862.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006019.mm.1 |
NM_182698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006019.mm.1 |
NM_182698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006019.mm.1 |
TC1100000755.mm.1 |
MTA TC |