Assay Details
| Gene Symbol: Myo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103171.1 | NP_001096641.1 | 32 - 33 | 7128 | 102 | |
| NM_010862.2 | NP_034992.2 | 33 - 34 | 7182 | 102 | ||
| NM_182698.2 | NP_874357.2 | 31 - 32 | 3366 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myo15
Genomic Map