Assay Details
| Gene Symbol: Myo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103171.1 | NP_001096641.1 | 40 - 41 | 8072 | 112 | |
| NM_010862.2 | NP_034992.2 | 41 - 42 | 8126 | 112 | ||
| NM_182698.2 | NP_874357.2 | 39 - 40 | 4310 | 112 | ||
| GenBank mRNA | AF053130.1 | - | 38 - 39 | 4278 | 112 | |
| AF144095.1 | - | 41 - 42 | 8126 | 112 | ||
| AY331132.1 | - | 40 - 41 | 8072 | 112 | ||
| AY331133.1 | - | 39 - 40 | 4310 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myo15
Genomic Map