Assay Details
| Gene Symbol: Myo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103171.1 | NP_001096641.1 | 46 - 47 | 8642 | 86 | |
| NM_010862.2 | NP_034992.2 | 47 - 48 | 8696 | 86 | ||
| NM_182698.2 | NP_874357.2 | 45 - 46 | 4880 | 86 | ||
| GenBank mRNA | AF053130.1 | - | 44 - 45 | 4848 | 86 | |
| AF144095.1 | - | 47 - 48 | 8696 | 86 | ||
| AY331132.1 | - | 46 - 47 | 8642 | 86 | ||
| AY331133.1 | - | 45 - 46 | 4880 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myo15
Genomic Map