Assay Details
| Gene Symbol: Myo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103171.1 | NP_001096641.1 | 52 - 53 | 9338 | 87 | |
| NM_010862.2 | NP_034992.2 | 53 - 54 | 9392 | 87 | ||
| NM_182698.2 | NP_874357.2 | 51 - 52 | 5576 | 87 | ||
| GenBank mRNA | AF144095.1 | - | 53 - 54 | 9392 | 87 | |
| AK030528.1 | - | 3 - 4 | 372 | 87 | ||
| AY331132.1 | - | 52 - 53 | 9338 | 87 | ||
| AY331133.1 | - | 51 - 52 | 5576 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myo15
Genomic Map