Assay Details
| Gene Symbol: Myo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103171.1 | NP_001096641.1 | 58 - 59 | 9867 | 94 | |
| NM_010862.2 | NP_034992.2 | 59 - 60 | 9921 | 94 | ||
| NM_182698.2 | NP_874357.2 | 57 - 58 | 6105 | 94 | ||
| GenBank mRNA | AF144095.1 | - | 59 - 60 | 9921 | 94 | |
| AK030528.1 | - | 10 - 11 | 952 | 94 | ||
| AY331132.1 | - | 58 - 59 | 9867 | 94 | ||
| AY331133.1 | - | 57 - 58 | 6105 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myo15
Genomic Map