Assay Details
| Gene Symbol: Olfr749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020288.2 | NP_064684.2 | 1 - 2 | 68 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_020288.2 transcripts.
Set Membership: |
|

Olfr749
Genomic Map