Assay Details
| Gene Symbol: Ucn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021290.2 | NP_067265.1 | 1 - 2 | 93 | 111 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_021290.2 transcripts.
Set Membership: |
|

Ucn
Genomic Map