Assay Details
| Gene Symbol: Gfer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023040.3 | NP_075527.2 | 1 - 2 | 327 | 104 | |
| GenBank mRNA | AB041561.1 | - | 1 - 2 | 244 | 104 | |
| AF148688.1 | - | 1 - 2 | 107 | 104 | ||
| AK146579.1 | - | 1 - 2 | 328 | 104 | ||
| BC023941.1 | - | 1 - 2 | 327 | 5342526 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF148688.1 transcripts.
Set Membership: |
|

Gfer
Genomic Map