Assay Details
| Gene Symbol: Tcea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159750.1 | NP_001153222.1 | 8 - 9 | 1046 | 98 | |
| NM_001159751.1 | NP_001153223.1 | 8 - 9 | 942 | 98 | ||
| NM_011541.4 | NP_035671.1 | 8 - 9 | 1049 | 98 | ||
| GenBank mRNA | AK136035.1 | - | 8 - 9 | 942 | 98 | |
| AK143593.1 | - | 8 - 9 | 907 | 98 | ||
| BC006022.1 | - | 8 - 9 | 924 | 3590733 | 98 | |
| BC061490.1 | - | 8 - 9 | 941 | 2582185 | 98 | |
| BC083127.1 | - | 8 - 9 | 912 | 6390364 | 98 | |
| M18209.1 | - | 7 - 8 | 845 | 98 | ||
| M18210.1 | - | 8 - 9 | 903 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tcea1
Genomic Map