Assay Details
| Gene Symbol: S100a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163525.2 | NP_001156997.1 | 1 - 2 | 51 | 89 | |
| GenBank mRNA | AA726094.1 | - | 1 - 2 | 44 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001163525.2 transcripts.
Set Membership: |
|

S100a14
Genomic Map