Assay Details
| Gene Symbol: Tmem29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164683.1 | NP_001158155.1 | 2 - 3 | 295 | 99 | |
| NM_001164684.1 | NP_001158156.1 | 2 - 3 | 264 | 99 | ||
| GenBank mRNA | AK018228.1 | - | 2 - 3 | 265 | 99 | |
| BY288280.1 | - | 2 - 3 | 295 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001164683.1, NM_001164684.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475870 | NM_001164686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475870 | AK012546.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475870 | BC058428.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475870 | NM_001164686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475870 | AK012546.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475870 | BC058428.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099279 | AK165152.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138117 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 434734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225326 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371371 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730318 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219954 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323573 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913532 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147031 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111164 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475870 | NM_001164686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475870 | AK012546.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475870 | BC058428.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475870 | NM_001164686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475870 | AK012546.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475870 | BC058428.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099279 | AK165152.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P475870 | NM_001164686.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P475870 | AK012546.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P475870 | BC058428.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P475870 | NM_001164686.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P475870 | AK012546.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P475870 | BC058428.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14858727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14709240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15013729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14812666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15153594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14898691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14861121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14861122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741757 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14858724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32741773 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10607249 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607248 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444708_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428930_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430413_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10607246 | - | - |
| Mouse Genome 430 2.0 Array | 1428930_at | - | - |
| Mouse Genome 430 2.0 Array | 1444708_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444708_at | - | - |
| Mouse Genome 430 2.0 Array | 1430413_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428930_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430413_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 382245_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 382245_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00019778.mm.1 | NM_001164683.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019778.mm.1 | NM_001164683.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019778.mm.1 | NM_001164684.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019778.mm.1 | NM_001164684.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019778.mm.1 | TC0X00003159.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00019777.mm.1 | NM_001164683.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019777.mm.1 | NM_001164683.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019777.mm.1 | NM_001164684.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019777.mm.1 | NM_001164684.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019777.mm.1 | TC0X00003159.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428930_PM_at | - | - |
| Mouse Expression Set 430 | 1430413_PM_at | - | - |
| Mouse Expression Set 430 | 1444708_at | - | - |
| Mouse Expression Set 430 | 1430413_at | - | - |
| Mouse Expression Set 430 | 1444708_PM_at | - | - |
| Mouse Expression Set 430 | 1428930_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444708_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430413_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444708_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430413_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428930_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428930_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240911 | - | - |

Tmem29
Genomic Map