| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900000664.mm.1 |
NM_001168516.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000664.mm.1 |
NM_001168516.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000664.mm.1 |
NM_001168517.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000664.mm.1 |
NM_001168517.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000664.mm.1 |
NM_027476.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000664.mm.1 |
NM_027476.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000664.mm.1 |
TC1900000047.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900000666.mm.1 |
NM_001168516.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000666.mm.1 |
NM_001168516.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000666.mm.1 |
NM_001168517.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000666.mm.1 |
NM_001168517.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000666.mm.1 |
NM_027476.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000666.mm.1 |
NM_027476.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000666.mm.1 |
TC1900000047.mm.1 |
MTA TC |