Assay Details
| Gene Symbol: Nup62-il4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171024.1 | NP_001164495.1 | 3 - 4 | 324 | 74 | |
| GenBank mRNA | AK133082.1 | - | 3 - 4 | 299 | 74 | |
| BY087056.1 | - | 3 - 4 | 325 | 74 | ||
| BY088421.1 | - | 3 - 4 | 325 | 74 | ||
| CJ047381.1 | - | 3 - 4 | 299 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nup62-il4i1
Genomic Map