Assay Details
| Gene Symbol: Csrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007792.4 | NP_031818.3 | 1 - 2 | 80 | 85 | |
| GenBank mRNA | AF037208.1 | - | 1 - 2 | 80 | 85 | |
| AK003328.1 | - | 1 - 2 | 79 | 85 | ||
| AK010626.1 | - | 1 - 2 | 81 | 85 | ||
| AK012647.1 | - | 1 - 2 | 78 | 85 | ||
| BC012663.1 | - | 1 - 2 | 65 | 4210909 | 85 | |
| BY333413.1 | - | 1 - 2 | 81 | 85 | ||
| CK031156.1 | - | 1 - 2 | 66 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF037208.1, AK003328.1, AK010626.1, AK012647.1, BC012663.1, NM_007792.4 transcripts.
Set Membership: |
|

Csrp2
Genomic Map