Assay Details
| Gene Symbol: Csrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007792.4 | NP_031818.3 | 4 - 5 | 488 | 97 | |
| GenBank mRNA | AF037208.1 | - | 4 - 5 | 488 | 97 | |
| AK003328.1 | - | 4 - 5 | 487 | 97 | ||
| AK010626.1 | - | 4 - 5 | 489 | 97 | ||
| AK012647.1 | - | 4 - 5 | 486 | 97 | ||
| BC012663.1 | - | 4 - 5 | 473 | 4210909 | 97 | |
| CK031156.1 | - | 4 - 5 | 474 | 97 | ||
| D88792.1 | - | 3 - 4 | 414 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csrp2
Genomic Map