Assay Details
| Gene Symbol: Cela2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007919.2 | NP_031945.1 | 1 - 2 | 62 | 102 | |
| GenBank mRNA | AK131813.1 | - | 1 - 2 | 63 | 102 | |
| AK160448.1 | - | 1 - 2 | 63 | 102 | ||
| BC026552.1 | - | 1 - 2 | 51 | 4990174 | 102 | |
| X04573.1 | - | 1 - 2 | 62 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC026552.1, NM_007919.2, X04573.1 transcripts.
Set Membership: |
|

Cela2a
Genomic Map