Assay Details
| Gene Symbol: Cela2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007919.2 | NP_031945.1 | 7 - 8 | 822 | 98 | |
| GenBank mRNA | BC026552.1 | - | 7 - 8 | 811 | 4990174 | 98 |
| X04573.1 | - | 7 - 8 | 822 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cela2a
Genomic Map