Assay Details
| Gene Symbol: Ifnab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008336.2 | NP_032362.2 | 1 | 252 | 105 | |
| GenBank mRNA | BC116862.1 | - | 1 | 252 | 40126181 | 105 |
| BC116864.1 | - | 1 | 252 | 40126183 | 105 | |
| Gene Symbol: Ifna6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206871.1 | NP_996754.1 | 1 | 211 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ifnab
Genomic Map