Assay Details
| Gene Symbol: P2ry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005013.2 | - | 2 - 3 | 474 | 102 | |
| AK017378.1 | - | 2 - 3 | 481 | 102 | ||
| AK155521.1 | - | 1 - 2 | 326 | 102 | ||
| AK155562.1 | - | 1 - 2 | 326 | 102 | ||
| BC006613.1 | - | 2 - 3 | 468 | 3967332 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005013.2, AK017378.1, AK155521.1, AK155562.1, BC006613.1 transcripts.
Set Membership: |
|

P2ry2
Genomic Map