Assay Details
| Gene Symbol: Aprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009698.2 | NP_033828.2 | 4 - 5 | 473 | 81 | |
| GenBank mRNA | AB033539.1 | - | 4 - 5 | 456 | 81 | |
| AK002350.1 | - | 4 - 5 | 426 | 81 | ||
| AK153201.1 | - | 4 - 5 | 438 | 81 | ||
| BC005667.1 | - | 4 - 5 | 411 | 3152733 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Aprt
Genomic Map