Assay Details
| Gene Symbol: Nmur1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010341.1 | NP_034471.1 | 1 - 2 | 832 | 58 | |
| GenBank mRNA | AF044602.1 | - | 1 - 2 | 832 | 58 | |
| BC120822.1 | - | 1 - 2 | 853 | 40129745 | 58 | |
| BC137775.1 | - | 1 - 2 | 853 | 8860794 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nmur1
Genomic Map