Assay Details
| Gene Symbol: Sec22b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011342.4 | NP_035472.1 | 3 - 4 | 462 | 86 | |
| GenBank mRNA | AK010363.1 | - | 3 - 4 | 471 | 86 | |
| AK017237.1 | - | 3 - 4 | 468 | 86 | ||
| AK034973.1 | - | 3 - 4 | 474 | 86 | ||
| AK037918.1 | - | 3 - 4 | 463 | 86 | ||
| AK088514.1 | - | 3 - 4 | 462 | 86 | ||
| AK089928.1 | - | 3 - 4 | 455 | 86 | ||
| AK134215.1 | - | 3 - 4 | 463 | 86 | ||
| AK190268.1 | - | 2 - 3 | 262 | 86 | ||
| BC009024.1 | - | 3 - 4 | 424 | 3481731 | 86 | |
| U91538.1 | - | 3 - 4 | 419 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sec22b
Genomic Map