Assay Details
| Gene Symbol: Cited2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010828.3 | NP_034958.2 | 1 - 2 | 229 | 77 | |
| GenBank mRNA | AK131664.1 | - | 1 - 2 | 232 | 77 | |
| AK133791.1 | - | 1 - 2 | 230 | 77 | ||
| AK183582.1 | - | 1 - 2 | 36 | 77 | ||
| BC057126.1 | - | 1 - 2 | 211 | 6415181 | 77 | |
| U86445.1 | - | 1 - 2 | 209 | 77 | ||
| Y15163.1 | - | 1 - 2 | 208 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK131664.1, AK133791.1, BC057126.1, NM_010828.3, U86445.1, Y15163.1 transcripts.
Set Membership: |
|

Cited2
Genomic Map