Assay Details
| Gene Symbol: Npy1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010934.4 | NP_035064.1 | 1 - 2 | 95 | 89 | |
| GenBank mRNA | AK132397.1 | - | 1 - 2 | 96 | 89 | |
| D63819.1 | - | 52 | 89 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK132397.1, D63819.1, NM_010934.4 transcripts.
Set Membership: |
|

Npy1r
Genomic Map