Assay Details
| Gene Symbol: Olfr263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010984.1 | NP_035114.1 | 1 | 445 | 101 | |
| GenBank mRNA | AF102516.1 | - | 1 | 256 | 101 | |
| BC116918.1 | - | 1 | 524 | 40126237 | 101 | |
| BC120698.1 | - | 1 | 524 | 40129621 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094787 | BC116918.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094787 | BC120698.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094787 | NM_010984.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094787 | AF102516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094787 | BC116918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094787 | BC120698.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094787 | NM_010984.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094787 | AF102516.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P244520 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P281950 | BC116918.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P281950 | BC120698.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P281950 | NM_010984.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P281950 | AF102516.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28871258 | - | - |
| Whole Mouse Genome G4122A | A_51_P244520 | - | - |
| Whole Mouse Genome G4122A | A_51_P281950 | BC116918.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281950 | BC120698.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281950 | NM_010984.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281950 | AF102516.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28871298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10876832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10699423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28871259 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10403900 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403902 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403901 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403899 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300001081.mm.1 | TC1300000160.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 767591 | - | - |
| ABI Mouse Genome Survey Microarray | 897904 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2060037 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2060037 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002060037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2592893 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2592893 | - | - |
| mouse-6 v1.1 (Illumina) | scl018341.1_205-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2060037 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl018341.1_205-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2060037 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2592893 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2592893 | - | - |

Olfr263
Genomic Map