Assay Details
| Gene Symbol: Uqcrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025407.2 | NP_079683.2 | 5 - 6 | 699 | 90 | |
| GenBank mRNA | AK002292.1 | - | 5 - 6 | 638 | 89 | |
| AK010553.1 | - | 5 - 6 | 639 | 90 | ||
| AK013128.1 | - | 5 - 6 | 639 | 90 | ||
| AK151764.1 | - | 5 - 6 | 641 | 90 | ||
| AK160337.1 | - | 5 - 6 | 646 | 90 | ||
| AK168218.1 | - | 5 - 6 | 641 | 90 | ||
| BC030064.1 | - | 5 - 6 | 636 | 1400271 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uqcrc1
Genomic Map