Assay Details
| Gene Symbol: Nanp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026086.2 | NP_080362.1 | 1 - 2 | 141 | 107 | |
| GenBank mRNA | AK005566.1 | - | 1 - 2 | 117 | 107 | |
| AK021344.1 | - | 1 - 2 | 128 | 107 | ||
| BC018527.1 | - | 1 - 2 | 112 | 3596532 | 107 | |
| BC083086.1 | - | 1 - 2 | 141 | 6515732 | 107 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nanp
Genomic Map