Assay Details
| Gene Symbol: Pdcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026850.4 | NP_081126.2 | 3 - 4 | 255 | 92 | |
| GenBank mRNA | AF110512.1 | - | 2 - 3 | 225 | 92 | |
| AK004334.1 | - | 3 - 4 | 250 | 92 | ||
| AK078372.1 | - | 3 - 4 | 256 | 92 | ||
| AK168509.1 | - | 3 - 4 | 296 | 92 | ||
| BC005601.1 | - | 3 - 4 | 276 | 3483084 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdcl3
Genomic Map