Assay Details
| Gene Symbol: Pdcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026850.4 | NP_081126.2 | 4 - 5 | 394 | 75 | |
| GenBank mRNA | AF110512.1 | - | 3 - 4 | 364 | 75 | |
| AK004334.1 | - | 4 - 5 | 389 | 75 | ||
| AK078372.1 | - | 4 - 5 | 395 | 75 | ||
| AK168509.1 | - | 4 - 5 | 435 | 75 | ||
| AK169572.1 | - | 4 - 5 | 385 | 75 | ||
| BC005601.1 | - | 4 - 5 | 415 | 3483084 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pdcl3
Genomic Map