Assay Details
| Gene Symbol: Ms4a6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027209.3 | NP_081485.2 | 6 - 7 | 770 | 98 | |
| GenBank mRNA | AF237909.1 | - | 6 - 7 | 770 | 98 | |
| AK007613.1 | - | 701 | 98 | |||
| AK075671.1 | - | 6 - 7 | 747 | 98 | ||
| AK172345.1 | - | 2518 | 98 | |||
| BC027425.1 | - | 6 - 7 | 747 | 3984299 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ms4a6b
Genomic Map