Assay Details
| Gene Symbol: Snrpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027246.1 | NP_081522.1 | 1 - 2 | 119 | 89 | |
| GenBank mRNA | AK008240.1 | - | 1 - 2 | 120 | 89 | |
| AK135835.1 | - | 1 - 2 | 115 | 89 | ||
| BC066015.1 | - | 1 - 2 | 82 | 89 | ||
| BC100499.1 | - | 1 - 2 | 145 | 89 | ||
| BQ748807.1 | - | 1 - 2 | 91 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008240.1, AK135835.1, BC100499.1, NM_027246.1 transcripts.
Set Membership: |
|

Snrpf
Genomic Map