Assay Details
| Gene Symbol: Enkur | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027728.2 | NP_082004.1 | 3 - 4 | 657 | 107 | |
| GenBank mRNA | AK017056.1 | - | 3 - 4 | 658 | 107 | |
| AY454124.1 | - | 3 - 4 | 655 | 107 | ||
| BC099519.1 | - | 3 - 4 | 556 | 30848009 | 107 | |
| BC132425.1 | - | 3 - 4 | 518 | 40130702 | 107 | |
| BC132451.1 | - | 3 - 4 | 518 | 40130728 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Enkur
Genomic Map