Assay Details
| Gene Symbol: Trnau1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027925.3 | NP_082201.2 | 3 - 4 | 387 | 101 | |
| GenBank mRNA | AK152986.1 | - | 3 - 4 | 336 | 101 | |
| BC005795.1 | - | 3 - 4 | 344 | 101 | ||
| BC048840.1 | - | 3 - 4 | 338 | 5008141 | 101 | |
| BC055454.1 | - | 3 - 4 | 350 | 4014366 | 101 | |
| BC099594.1 | - | 1 - 2 | 72 | 101 | ||
| CJ166630.1 | - | 3 - 4 | 350 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trnau1ap
Genomic Map