Assay Details
| Gene Symbol: Trnau1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027925.3 | NP_082201.2 | 7 - 8 | 850 | 75 | |
| GenBank mRNA | AK003534.1 | - | 6 - 7 | 1513 | 75 | |
| AK152986.1 | - | 7 - 8 | 799 | 75 | ||
| BC048840.1 | - | 7 - 8 | 801 | 5008141 | 75 | |
| BC055454.1 | - | 7 - 8 | 813 | 4014366 | 75 | |
| BC099594.1 | - | 5 - 6 | 535 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trnau1ap
Genomic Map