Assay Details
| Gene Symbol: Gjc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080454.4 | NP_536702.3 | 1 - 2 | 55 | 82 | |
| GenBank mRNA | AK046609.1 | - | 1 - 2 | 56 | 82 | |
| AY394498.1 | - | 1 - 2 | 55 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046609.1, AY394498.1, NM_080454.4 transcripts.
Set Membership: |
|

Gjc2
Genomic Map