Assay Details
| Gene Symbol: H1foo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138311.2 | NP_612184.1 | 1 - 2 | 149 | 99 | |
| GenBank mRNA | AK135944.1 | - | 1 - 2 | 153 | 99 | |
| AK162137.1 | - | 1 - 2 | 128 | 99 | ||
| AK162154.1 | - | 1 - 2 | 146 | 99 | ||
| AY007195.1 | - | 1 - 2 | 149 | 99 | ||
| BC137916.1 | - | 1 - 2 | 145 | 8860935 | 99 | |
| BC145046.1 | - | 1 - 2 | 145 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

H1foo
Genomic Map